Learn More
Glass Expansion Quartz Torch with single slot for Avio 200 (N0790131)
SDP
Catalog No. 501907255
Encompass_Preferred
Quartz Torch, 1-Slot demountable quartz torch standard for PerkinElmer Avio 200 and 500 Series ICP-OES (N0790131).Standard torch for routine aqueous applications, but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7255 Supplier Glass Expansion Supplier No. 318083776
Quartz Torch, 1-Slot demountable quartz torch standard for PerkinElmer Avio 200 and 500 Series ICP-OES (N0790131).Standard torch for routine aqueous applications, but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001