Learn More
Glass Expansion Quartz Torch without slot for Optima 8x00
SDP
Catalog No. 501907193
Encompass_Preferred
Quartz Torch, 0-Slot demountable quartz torch standard for PerkinElmer Optima 8x00 Series ICP-OES (N0780133).Recommended for organic solvent applications, but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7193 Supplier Glass Expansion Supplier No. 318083254
Quartz Torch, 0-Slot demountable quartz torch standard for PerkinElmer Optima 8x00 Series ICP-OES (N0780133).Recommended for organic solvent applications, but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001