Learn More
Glass Expansion Quartz Torch for SpectroBlue EOP
SDP
Catalog No. 501907217
Encompass_Preferred
Quartz Torch, semi-demountable torch for Spectro (Ametek) SpectroBlue EOP ICP-OES (48205043).Suitable for routine aqueous and organic applications, but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7217 Supplier Glass Expansion Supplier No. 318083394
Quartz Torch, semi-demountable torch for Spectro (Ametek) SpectroBlue EOP ICP-OES (48205043).Suitable for routine aqueous and organic applications, but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001