Learn More
Glass Expansion Standard quartz torch body for Thermo Q/RQ/TQ Series ICP-MS (1230790, 1500101201)
SDP
Catalog No. 501694609
Encompass_Preferred
Quartz Torch, standard, no injector, body only, for aqueous and organic solvent analysis with Thermo Q/RQ/TQ Sereis ICP-MS (1230790). Standard quartz torch suitable for routine aqueous and organic applications but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-169-4609 Supplier Glass Expansion Supplier No. 318083357
Quartz Torch, standard, no injector, body only, for aqueous and organic solvent analysis with Thermo Q/RQ/TQ Sereis ICP-MS (1230790). Standard quartz torch suitable for routine aqueous and organic applications but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001