Learn More
Perkin Elmer US LLC 1-Slot Demountable Quartz Torch for Optima 3000/3000 SCR/3x00 RL
Catalog No. 509043687
Encompass_Preferred
This demountable 1-slot torch design uses one-piece quartz tubing for plasma and auxiliary gas flow and is the standard torch for aqueous solutions.
Catalog No. 50-904-3687 Supplier Perkin Elmer US LLC Supplier No. N0690568
This demountable 1-slot torch design uses one-piece quartz tubing for plasma and auxiliary gas flow and is the standard torch for aqueous solutions.