Learn More
Glass Expansion Standard quartz torch body for iCAP 6000/7000 Radial, Glass Expansion, body only, no injector (842312051741, BRE0019945)
SDP
Catalog No. 501694685
Encompass_Preferred
Quartz Torch, standard, no injector, body only, for aqueous and organic solvent analysis with Thermo 6000/7000 Radial (EMT) Sereis ICP-OES (842312051741). Standard quartz torch suitable for routine aqueous and organic applications but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-169-4685 Supplier Glass Expansion Supplier No. 318082810
Quartz Torch, standard, no injector, body only, for aqueous and organic solvent analysis with Thermo 6000/7000 Radial (EMT) Sereis ICP-OES (842312051741). Standard quartz torch suitable for routine aqueous and organic applications but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001