Learn More
Glass Expansion Quartz Torch, single-piece, with ball joint, RA/FS13 & 2.3mm ID Injector for Agilent (Varian) 700-ES, Liberty and Vista Series ICP-OES (7910050700).
SDP
Catalog No. 502703167
Encompass_Preferred
Quartz Torch, single-piece, RA/FS13 & 2.3 mm ID injector, for Agilent (Varian) 700-ES, Liberty and Vista Axial Series ICP-OES (7910050700). Recommended for routine environmental sample analysis with high TDS but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3167 Supplier Glass Expansion Supplier No. 308080540
Quartz Torch, single-piece, RA/FS13 & 2.3 mm ID injector, for Agilent (Varian) 700-ES, Liberty and Vista Axial Series ICP-OES (7910050700). Recommended for routine environmental sample analysis with high TDS but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001