Learn More
Agilent Technologies ICP-MS Quartz Torch, Single-Piece, 2.5 mm ID, 1 ea
Catalog No. NC2742311
Encompass
This single-piece quartz torch is designed for use in Agilent 7700, 7800, 7850, 7900, 8800, and 8900 ICP-MS systems. It is the standard recommended torch for routine analysis of aqueous samples. The unit consists of three concentric quartz tubes that manage the plasma, auxiliary, and nebulizer gas streams.
  • Compatible with Agilent 7700, 7800, 7850, 7900, 8800, and 8900 ICP-MS series
  • Features a 2.5 mm inner diameter quartz injector
  • Ideal for most routine aqueous sample applications
  • Not suitable for use with volatile organic solvents or samples containing hydrofluoric acid
  • Requires complete drying before installation and plasma ignition
Catalog No. NC2742311 Supplier Agilent Technologies Supplier No. G328080053
This single-piece quartz torch is designed for use in Agilent 7700, 7800, 7850, 7900, 8800, and 8900 ICP-MS systems. It is the standard recommended torch for routine analysis of aqueous samples. The unit consists of three concentric quartz tubes that manage the plasma, auxiliary, and nebulizer gas streams.
  • Compatible with Agilent 7700, 7800, 7850, 7900, 8800, and 8900 ICP-MS series
  • Features a 2.5 mm inner diameter quartz injector
  • Ideal for most routine aqueous sample applications
  • Not suitable for use with volatile organic solvents or samples containing hydrofluoric acid
  • Requires complete drying before installation and plasma ignition