Learn More
Glass Expansion Quartz Torch for Wear Metals, 1.8mm injector with gas fittings for Spectroflame
SDP
Catalog No. 5000556650
Encompass_Preferred
Quartz Torch for Wear Metals, 1.8mm injector with gas fittings for Spectroflame Sourced from Glass Expansion
Catalog No. 50-005-56650 Supplier Glass Expansion Supplier No. 308073669
Quartz Torch for Wear Metals, 1.8mm injector with gas fittings for Spectroflame Sourced from Glass Expansion