Learn More
Mti Corporation QUARTZ TUBE 1"O.D. X 17" L
SDP
Catalog No. NC1964242
Encompass
Quartz Tube: 25 OD x 20.5 IDx 450 L mm (1" O.D. x 17.7" L) for DIY Tube Furnace - EQ-QZTube-25GE-450
Catalog No. NC1964242 Supplier Mti Corporation Supplier No. TQ25D450
Quartz Tube: 25 OD x 20.5 IDx 450 L mm (1" O.D. x 17.7" L) for DIY Tube Furnace - EQ-QZTube-25GE-450