Learn More
Glass Expansion Quartz Tube Set for 5100 RV Demountable Torch
SDP
Catalog No. 501907230
Encompass_Preferred
Semi Demountable Torch (SDT) Body, no injector. Compatible with Agilent 5100 RV ICP-OES.Semi-demountable quartz torch body suitable for routine aqueous and organic applications but not inert to samples containing hydrofluoric acid (HF). Optional injectors include 1.8 mm Tapered Quartz Injector (501907232) for most routine aqueous and organic applications. The 2.4 mm Tapered Quartz Injector (31-808-3584) for most high dissolved solids applications, 1.8 mm Tapered Alumina Injector (501907235) for routine aqueous with HF, and the 2.4 mm Tapered Alumina Injector (501907251) for HF and high dissolved solids applications. For volatile organic solvent applications the 0.8 mm Tapered Quartz Injector (31-808-3738). The 1.4 mm Tapered Quartz Injector (31-808-3583) for less volatile organic solvents, 1.5 mm Tapered Alumina Injector (31-808-3727) for use with aqueous, including HF, and organic solvent-based matrices.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7230 Supplier Glass Expansion Supplier No. 318083556
Semi Demountable Torch (SDT) Body, no injector. Compatible with Agilent 5100 RV ICP-OES.Semi-demountable quartz torch body suitable for routine aqueous and organic applications but not inert to samples containing hydrofluoric acid (HF). Optional injectors include 1.8 mm Tapered Quartz Injector (501907232) for most routine aqueous and organic applications. The 2.4 mm Tapered Quartz Injector (31-808-3584) for most high dissolved solids applications, 1.8 mm Tapered Alumina Injector (501907235) for routine aqueous with HF, and the 2.4 mm Tapered Alumina Injector (501907251) for HF and high dissolved solids applications. For volatile organic solvent applications the 0.8 mm Tapered Quartz Injector (31-808-3738). The 1.4 mm Tapered Quartz Injector (31-808-3583) for less volatile organic solvents, 1.5 mm Tapered Alumina Injector (31-808-3727) for use with aqueous, including HF, and organic solvent-based matrices.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001