Learn More
Glass Expansion Quartz Tube Set for 5100 SVDV/VDV Demountable Torch (G8010-60263)
SDP
Catalog No. 501907231
Encompass_Preferred
Semi Demountable Torch (SDT) Body, no injector. Compatible with Agilent 5100 SVDV/VDV ICP-OES.Semi-demountable quartz torch body suitable for routine aqueous & organic applications, but not inert to samples containing hydrofluoric acid (HF). Optional injectors include 1.8 mm Tapered Quartz Injector (501907232) for most routine aqueous & organic applications. The 2.4 mm Tapered Quartz Injector (31-808-3584) for most high dissolved solids applications, 1.8 mm Tapered Alumina Injector (501907235) for routine aqueous with HF, & the 2.4 mm Tapered Alumina Injector (501907251) for HF & high dissolved solids applications. For volatile organic solvent applications the 0.8 mm Tapered Quartz Injector (31-808-3738). The 1.4 mm Tapered Quartz Injector (31-808-3583) for less volatile organic solvents, 1.5 mm Tapered Alumina Injector (31-808-3727) for use with aqueous, including HF, and organic solvent-based matrices.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7231 Supplier Glass Expansion Supplier No. 318083557
Semi Demountable Torch (SDT) Body, no injector. Compatible with Agilent 5100 SVDV/VDV ICP-OES.Semi-demountable quartz torch body suitable for routine aqueous & organic applications, but not inert to samples containing hydrofluoric acid (HF). Optional injectors include 1.8 mm Tapered Quartz Injector (501907232) for most routine aqueous & organic applications. The 2.4 mm Tapered Quartz Injector (31-808-3584) for most high dissolved solids applications, 1.8 mm Tapered Alumina Injector (501907235) for routine aqueous with HF, & the 2.4 mm Tapered Alumina Injector (501907251) for HF & high dissolved solids applications. For volatile organic solvent applications the 0.8 mm Tapered Quartz Injector (31-808-3738). The 1.4 mm Tapered Quartz Injector (31-808-3583) for less volatile organic solvents, 1.5 mm Tapered Alumina Injector (31-808-3727) for use with aqueous, including HF, and organic solvent-based matrices.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001