Learn More
Electron Microscopy Sciences Quasar Lighting System, Kit
SDP
Catalog No. 5032964
Encompass_Preferred
Custom fitting patent pending LED Lighting attachment for Optivision Head Band Magnifiers reduces shadows and increases visibility in any environment and lights up holes and deep cavities. Super long batteries life with light remaining bright after 16 hours of straight use and lasts up to 96 hours solid on a single set of batteries. Over 100,000 hour bulb life expectanct. Kit includes lighting system frame, retro fit attachment pins, longer screw for OptiLOUPE fitted visors, 2 AA batteries and velcro strap.
Catalog No. 50-329-64 Supplier Electron Microscopy Sciences Supplier No. 6810770
Custom fitting patent pending LED Lighting attachment for Optivision Head Band Magnifiers reduces shadows and increases visibility in any environment and lights up holes and deep cavities. Super long batteries life with light remaining bright after 16 hours of straight use and lasts up to 96 hours solid on a single set of batteries. Over 100,000 hour bulb life expectanct. Kit includes lighting system frame, retro fit attachment pins, longer screw for OptiLOUPE fitted visors, 2 AA batteries and velcro strap.