Learn More
Cayman Chemical QuasIa amara QuasIn 10mg
SDP
Catalog No. 502946298
Encompass_Preferred
A quassinoid with diverse biological activities; active against chloroquine-sensitive and -resistant strains of P. falciparum (IC50 = 0.15 µM for both); has antifeedant and insecticidal activity against the diamondback moth when applied to dietary cabbage leaf discs at 16, 31.9, or 63.7 UG/cm2; increases viability of SH-SY5Y cells in a model of hydrogen peroxide-induced neuronal injury at 25, 50, or 100 µM
Catalog No. 50-294-6298 Supplier Cayman Chemical Supplier No. 3438210 MG
A quassinoid with diverse biological activities; active against chloroquine-sensitive and -resistant strains of P. falciparum (IC50 = 0.15 µM for both); has antifeedant and insecticidal activity against the diamondback moth when applied to dietary cabbage leaf discs at 16, 31.9, or 63.7 UG/cm2; increases viability of SH-SY5Y cells in a model of hydrogen peroxide-induced neuronal injury at 25, 50, or 100 µM