Learn More
MSC Betco Disinfectant, 5 Minute Contact Time, Alkaline, 1:256 Dilution
Catalog No. 502836010
Encompass
Quat-Stat 5 is a hospital disinfection product with a 5-minute contact time. This one-step registered cleaner and disinfectant effectively cleans, disinfects, and deodorizes a variety of surfaces. It is also effective against SARS-Related Coronavirus SARS-CoV-2 in 1 minute and is approved for use in electrostatic sprayers.
  • Five-minute dwell time
  • Effective yet economical with a 1:256 dilution
  • Efficacy against Norovirus
  • Saves labor cost with a 5-minute dwell time
  • Can be used through an auto-scrubber
  • Cleans, disinfects, and deodorizes
  • Effective against SARS-Related Coronavirus SARS-CoV-2 in 1 minute
  • Kills HIV, HBV, and HCV on pre-cleaned environmental surfaces/objects previously soiled with blood/body fluids
Catalog No. 50-283-6010 Supplier MSC Supplier No. 73006868
Quat-Stat 5 is a hospital disinfection product with a 5-minute contact time. This one-step registered cleaner and disinfectant effectively cleans, disinfects, and deodorizes a variety of surfaces. It is also effective against SARS-Related Coronavirus SARS-CoV-2 in 1 minute and is approved for use in electrostatic sprayers.
  • Five-minute dwell time
  • Effective yet economical with a 1:256 dilution
  • Efficacy against Norovirus
  • Saves labor cost with a 5-minute dwell time
  • Can be used through an auto-scrubber
  • Cleans, disinfects, and deodorizes
  • Effective against SARS-Related Coronavirus SARS-CoV-2 in 1 minute
  • Kills HIV, HBV, and HCV on pre-cleaned environmental surfaces/objects previously soiled with blood/body fluids