Learn More
Grainger Quat Test Kit 7 1/2 in L 0 to 400 ppm
Catalog No. 502753499
Encompass_Preferred
The Hydrion Quat Test Kit 0 to 400 ppm is a test kit used to measure quaternary ammonium compounds in solution. It contains 100 test papers and a color chart to determine quaternary ammonium concentration between 0-400 ppm. The test papers change color when dipped in solution, with the color matched to the chart to find the corresponding concentration. This allows quick on-site testing of quaternary ammonium solutions. The kit is portable, requires no additional reagents or equipment, and is EPA compliant. It is suitable for testing cleaners, sanitizers and disinfectants that contain quaternary ammonium compounds.
Catalog No. 50-275-3499 Supplier Grainger Supplier No. 14G846
The Hydrion Quat Test Kit 0 to 400 ppm is a test kit used to measure quaternary ammonium compounds in solution. It contains 100 test papers and a color chart to determine quaternary ammonium concentration between 0-400 ppm. The test papers change color when dipped in solution, with the color matched to the chart to find the corresponding concentration. This allows quick on-site testing of quaternary ammonium solutions. The kit is portable, requires no additional reagents or equipment, and is EPA compliant. It is suitable for testing cleaners, sanitizers and disinfectants that contain quaternary ammonium compounds.