Learn More
Pharmacal Research Laboratories QUATRICIDE PV 1 GALLON
SDP
Catalog No. NC0282465
Encompass
QUATRICIDE PV 1 GALLON. Quatricide� PV is a disinfectant/detergent that will clean and disinfect in one step. Quatricide� PV is Fungicidal, Bactericidal and Virucidal (against certain listed enveloped and non-enveloped viruses). Efficacy is determined by the AOAC Use-Dilution test method on hard inanimate surfaces, modified in the presence of 400 ppm synthetic hard water (calculated as CaCo3) plus 5% organic serum. This product does not contain any perfumes or fragrances. Quatricide� PV is used at a dilution of 1:64 or 2 ounces per gallon of water
This item is not returnable. View return policy
Catalog No. NC0282465 Supplier Pharmacal Research Laboratories Supplier No. 68018F
QUATRICIDE PV 1 GALLON. Quatricide� PV is a disinfectant/detergent that will clean and disinfect in one step. Quatricide� PV is Fungicidal, Bactericidal and Virucidal (against certain listed enveloped and non-enveloped viruses). Efficacy is determined by the AOAC Use-Dilution test method on hard inanimate surfaces, modified in the presence of 400 ppm synthetic hard water (calculated as CaCo3) plus 5% organic serum. This product does not contain any perfumes or fragrances. Quatricide� PV is used at a dilution of 1:64 or 2 ounces per gallon of water