Learn More
Pharmacal Research Laboratories QUATRICIDE PV-15 DISINFECTANT
SDP
Catalog No. NC1554689
Encompass
Quatricide® PV-15 is a disinfectant/detergent that will clean, disinfect and deodorize in one step. Quatricide® PV-15 is Fungicidal, Bactericidal and Virucidal (against certain listed enveloped and non-enveloped viruses). Efficacy is determined by the AOA
This item is not returnable. View return policy
Catalog No. NC1554689 Supplier Pharmacal Research Laboratories Supplier No. 64020
Quatricide® PV-15 is a disinfectant/detergent that will clean, disinfect and deodorize in one step. Quatricide® PV-15 is Fungicidal, Bactericidal and Virucidal (against certain listed enveloped and non-enveloped viruses). Efficacy is determined by the AOA