Learn More
Pharmacal Research Laboratories QUATRICIDE PV15 1 GALLON, 4/CS
SDP
Catalog No. NC9371434
Encompass
Quatricide® PV-15 is a disinfectant/detergent that will clean and disinfect in one step. Quatricide® PV-15 is Fungicidal, Bactericidal and Virucidal (against certain listed enveloped and non-enveloped viruses). Efficacy is determined by the AOAC Use-Dilution test method on hard inanimate surfaces, modified in the presence of 400 ppm synthetic hard water (calculated as CaCo3) plus 5% organic serum. This product does not contain any perfumes or fragrances. Quatricide® PV-15 is used at a dilution of 1:256 or 1/2 ounces per gallon of water.
This item is not returnable. View return policy
Catalog No. NC9371434 Supplier Pharmacal Research Laboratories Supplier No. 64020F
Quatricide® PV-15 is a disinfectant/detergent that will clean and disinfect in one step. Quatricide® PV-15 is Fungicidal, Bactericidal and Virucidal (against certain listed enveloped and non-enveloped viruses). Efficacy is determined by the AOAC Use-Dilution test method on hard inanimate surfaces, modified in the presence of 400 ppm synthetic hard water (calculated as CaCo3) plus 5% organic serum. This product does not contain any perfumes or fragrances. Quatricide® PV-15 is used at a dilution of 1:256 or 1/2 ounces per gallon of water.