Learn More
Pharmacal Research Laboratories QUATRICIDE TB 32OZ 12/CS
SDP
Catalog No. NC9816281
Encompass
Quatricide TB, 32 oz, Boiling Point: 200 deg.F, Specific Gravity: 1.004, pH: 11.7, Appearance: Clear Liquid, Solubility in Water at 1 Atm and 25 deg.C
This item is not returnable. View return policy
Catalog No. NC9816281 Supplier Pharmacal Research Laboratories Supplier No. 63032F
Quatricide TB, 32 oz, Boiling Point: 200 deg.F, Specific Gravity: 1.004, pH: 11.7, Appearance: Clear Liquid, Solubility in Water at 1 Atm and 25 deg.C