Learn More
Diagnostic Biosystems QUATRO SOL II QuatroSol II 10X Tris EDTA Buffer For Heat Induced Epitope Recovery, pH 9.0 kit/200 Runs
Catalog No. 502407360
Encompass_Preferred
QuatroSol I 10X Citrate Buffer For Heat Induced Epitope Recovery, pH 6.0 kit is designed for use during the heat induced epitope retrieval (HIER) step prior to immunohistochemistry on formalin-fixed paraffin embedded tissue sections. The use of this buffer in combination with heat (often by microwave, water bath, or pressure cooker) has been shown to restore the antigenicity of proteins modified during the formalin fixation of tissue. This buffer is supplied as a 10X stock solution.\n\n 
Catalog No. 50-240-7360 Supplier Diagnostic Biosystems Supplier No. K086
QuatroSol I 10X Citrate Buffer For Heat Induced Epitope Recovery, pH 6.0 kit is designed for use during the heat induced epitope retrieval (HIER) step prior to immunohistochemistry on formalin-fixed paraffin embedded tissue sections. The use of this buffer in combination with heat (often by microwave, water bath, or pressure cooker) has been shown to restore the antigenicity of proteins modified during the formalin fixation of tissue. This buffer is supplied as a 10X stock solution.\n\n 