Learn More
Accuflow Systems Quattroflow Diaphragm Pump, Stainless Steel Chamber, 0.83-83 Lpm, 230 V
Supplier:  Accuflow Systems QF44HTACUEGN230
Encompass_Preferred
The QF4400HT series quaternary diaphragm pump delivers gentle, seal-free transfer of shear-sensitive aqueous and biological fluids. The HT compact configuration integrates the pump chamber, drive, motor, and controller into a single unit. This multi-use configuration features an SS316L pump chamber suitable for CIP/SIP and autoclave processes, while single-use options use machined polypropylene chambers for disposable applications.
  • Quaternary (four-piston) diaphragm design for gentle fluid handling.
  • SS316L pump chamber, compatible with CIP/SIP and autoclaving.
  • Flow range 0.83-83 Lpm (50-5,000 L/h) for wide turndown control.
  • Motor voltage 3x200-240 V for the -230 configuration.
  • No mechanical shaft seal or wetted rotating parts, ensuring total product containment.
  • HT compact integrated drive and controller for a space-saving footprint.
Catalog No. NC3884606
Add to Cart
Request a Quote