Learn More
Accuflow Systems QUATTROFLOW MODEL 150-SU
Catalog No. NC2587838
Encompass_Preferred
  • Compact, 4-piston diaphragm pump
  • Single-use wetted path with polypropylene chamber
  • Gentle handling for sensitive bioproducts
  • Integrated controller and digital keypad
  • Self-priming and dry-running capabilities
  • Suitable for filtration, chromatography, and feed pumping
  • Compact design for laboratory and R&D use
  • Maximum discharge pressure: 4 bar (58 psi)
  • Flow rate range: 1 to 180 L/hr
  • Voltage: 230 V (110 V optional)
Catalog No. NC2587838 Supplier Accuflow Systems Supplier No. AFSQF15DISPP3
  • Compact, 4-piston diaphragm pump
  • Single-use wetted path with polypropylene chamber
  • Gentle handling for sensitive bioproducts
  • Integrated controller and digital keypad
  • Self-priming and dry-running capabilities
  • Suitable for filtration, chromatography, and feed pumping
  • Compact design for laboratory and R&D use
  • Maximum discharge pressure: 4 bar (58 psi)
  • Flow rate range: 1 to 180 L/hr
  • Voltage: 230 V (110 V optional)