Learn More
Accuflow Systems QUATTROFLOW PUMP (CHAMBER KIT)
Catalog No. 501142826
Encompass_Preferred
DISPOSABLE CHAMBER CONSTRUCTED FROM POLYPROPYLENE/SANTOPRENE/EPDM USP CLASS VI STANDARDS. FITTED WITH 1/4 INCH CLAMP CONNECTIONS (OPTIONS AVAILABLE). DOCUMENTATION INCLUDES USP CLASS VI DECLARATIONS AND CERTIFICATE OF CONFORMANCE. EACH CHAMBER IS LABELED AND DOUBLE BAGGED.
Catalog No. 50-114-2826 Supplier Accuflow Systems Supplier No. QF15DISPP3
DISPOSABLE CHAMBER CONSTRUCTED FROM POLYPROPYLENE/SANTOPRENE/EPDM USP CLASS VI STANDARDS. FITTED WITH 1/4 INCH CLAMP CONNECTIONS (OPTIONS AVAILABLE). DOCUMENTATION INCLUDES USP CLASS VI DECLARATIONS AND CERTIFICATE OF CONFORMANCE. EACH CHAMBER IS LABELED AND DOUBLE BAGGED.