Learn More
Enzo Life Sciences Quazinone (10mg). CAS: 70018-51-8
SDP
Supplier:  Enzo Life Sciences BMLPD1700010
Encompass_Preferred
Selective inhibitor of cGMP-inhibited phosphodiesterase (PDE III, IC50=0.6µM). Alternative name: Ro 13-6438, (R)-6-Chloro-1,5-dihydro-3-methyl- imidazo[2,1-b]quinazolin-2-one. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (3mg/ml) or 100% ethanol (1mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-1385