Learn More
Waters Corp QuEChERS dSPE Tube, 1200 mg MgSO4, 400 mg PSA, 400 mg C18, 15 mL
Catalog No. 502392407 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This dSPE tube is designed for the clean-up step in QuEChERS sample preparation. It contains a blend of 1200 mg MgSO4, 400 mg PSA, and 400 mg C18, making it suitable for cleaning fatty produce and cereals. QuEChERS methods are commonly used for multi-pesticide residue analyses in food applications.
  • Designed for QuEChERS sample preparation technique.
  • Contains 1200 mg MgSO4, 400 mg PSA, and 400 mg C18 for sample clean-up.
  • Suitable for cleaning fatty produce and cereals in multi-pesticide residue analyses.
  • Supplied in a 15 mL tube.
  • Available in packages of 50 units.
  • Features a blue cap.
  • Supports AOAC methods.
Catalog No. 50-239-2407 Supplier Waters Corp Supplier No. 186008073
This dSPE tube is designed for the clean-up step in QuEChERS sample preparation. It contains a blend of 1200 mg MgSO4, 400 mg PSA, and 400 mg C18, making it suitable for cleaning fatty produce and cereals. QuEChERS methods are commonly used for multi-pesticide residue analyses in food applications.
  • Designed for QuEChERS sample preparation technique.
  • Contains 1200 mg MgSO4, 400 mg PSA, and 400 mg C18 for sample clean-up.
  • Suitable for cleaning fatty produce and cereals in multi-pesticide residue analyses.
  • Supplied in a 15 mL tube.
  • Available in packages of 50 units.
  • Features a blue cap.
  • Supports AOAC methods.