Learn More
Waters Corp QuEChERS Tube, 1200 mg MgSO4, 400 mg PSA, 15 mL, 50/pk
Catalog No. 502390638 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This clean-up tube is designed for the first step of QuEChERS sample preparation. It contains a combination of PSA, C18, and GCB for effective clean-up of fatty and pigmented samples, commonly encountered in multi-pesticide residue analyses in food.
  • Used for extract clean-up in QuEChERS sample preparation.
  • Contains 1200 mg MgSO4, 400 mg PSA, 400 mg C18, and 400 mg GCB for effective clean-up.
  • Designed for fatty and pigmented samples.
  • Supports multi-pesticide residue analyses in food.
  • Features a 15 mL tube size.
  • Utilizes QuEChERS and dispersive SPE techniques.
  • Adheres to AOAC methods.
  • Supplied as 50 tubes per package.
Catalog No. 50-239-0638 Supplier Waters Corp Supplier No. 186008074
This clean-up tube is designed for the first step of QuEChERS sample preparation. It contains a combination of PSA, C18, and GCB for effective clean-up of fatty and pigmented samples, commonly encountered in multi-pesticide residue analyses in food.
  • Used for extract clean-up in QuEChERS sample preparation.
  • Contains 1200 mg MgSO4, 400 mg PSA, 400 mg C18, and 400 mg GCB for effective clean-up.
  • Designed for fatty and pigmented samples.
  • Supports multi-pesticide residue analyses in food.
  • Features a 15 mL tube size.
  • Utilizes QuEChERS and dispersive SPE techniques.
  • Adheres to AOAC methods.
  • Supplied as 50 tubes per package.