Learn More
Waters Corp QuEChERS Tube, 1200 mg MgSO4/400 mg PSA, 15 mL
Catalog No. 502391579 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This clean-up tube is designed for use with extracts from step 1 of QuEChERS sample preparation. It contains d-SPE components, including PSA, for efficient cleaning of high water content samples like fruits and vegetables. QuEChERS methods are primarily employed for multi-pesticide residue analyses in food.
  • Contains PSA for cleaning high water content samples.
  • Used in QuEChERS methods for multi-pesticide residue analyses in food.
  • Supplied as a 50-pack of tubes.
Catalog No. 50-239-1579 Supplier Waters Corp Supplier No. 186008072
This clean-up tube is designed for use with extracts from step 1 of QuEChERS sample preparation. It contains d-SPE components, including PSA, for efficient cleaning of high water content samples like fruits and vegetables. QuEChERS methods are primarily employed for multi-pesticide residue analyses in food.
  • Contains PSA for cleaning high water content samples.
  • Used in QuEChERS methods for multi-pesticide residue analyses in food.
  • Supplied as a 50-pack of tubes.