Learn More
Waters Corp QuEChERS Tube, 150 mg MgSO4, PSA/C18/GCB, 2 mL
Catalog No. 502392687 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This cleanup tube is designed for use in step 1 of QuEChERS sample preparation. The d-SPE tube contains PSA, C18, and GCB for cleaning up samples such as spices and tea. QuEChERS methods are utilized for multi-pesticide residue analyses in food.
  • Designed for QuEChERS sample preparation.
  • Contains 150 mg MgSO4.
  • Features a d-SPE tube with 25 mg PSA, 25 mg C18, and 7 mg GCB.
  • Suitable for cleaning up samples such as spices and tea.
  • Utilized for multi-pesticide residue analyses in food.
  • Features a 2 mL tube format.
  • Supplied as 100 tubes per package.
Catalog No. 50-239-2687 Supplier Waters Corp Supplier No. 186008071
This cleanup tube is designed for use in step 1 of QuEChERS sample preparation. The d-SPE tube contains PSA, C18, and GCB for cleaning up samples such as spices and tea. QuEChERS methods are utilized for multi-pesticide residue analyses in food.
  • Designed for QuEChERS sample preparation.
  • Contains 150 mg MgSO4.
  • Features a d-SPE tube with 25 mg PSA, 25 mg C18, and 7 mg GCB.
  • Suitable for cleaning up samples such as spices and tea.
  • Utilized for multi-pesticide residue analyses in food.
  • Features a 2 mL tube format.
  • Supplied as 100 tubes per package.