Learn More
Waters Corp QuEChERS d-SPE Tube, 2 mL, 100 pk
Supplier: Waters Corp 186008076
This clean-up tube is designed for use with extracts from step 1 of QuEChERS sample preparation. The d-SPE tube contains PSA and GCB for the removal of chlorophyll and carotenoids, making it suitable for multi-pesticide residue analyses in food.
- Designed for use with extracts from step 1 of QuEChERS sample preparation.
- Contains PSA and GCB for the removal of chlorophyll and carotenoids.
- Utilized for multi-pesticide residue analyses in food.
- Packaged as 100 tubes.
- Features a 2 mL tube size.
- Supports QuEChERS and dispersive SPE techniques.