Learn More
Waters Corp QuEChERS Sample Cleanup, 150 mg MgSO4, 50 mg C18, 2 mL Tube
Catalog No. 502391664 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This clean-up tube is designed for use in the first step of QuEChERS sample preparation. It is a d-SPE tube containing C18, specifically formulated for the analysis of acidic pesticides. QuEChERS methods are widely used for multi-pesticide residue analyses in food samples.
  • Used for extract from step 1 in QuEChERS sample preparation.
  • d-SPE tube contains C18 for acidic pesticide analysis.
  • Utilized for multi-pesticide residue analyses in food.
  • Contains 150 mg MgSO4 and 50 mg C18.
  • Tube size of 2 mL.
  • Suitable for AOAC methods.
  • Supports QuEChERS and Dispersive SPE techniques.
Catalog No. 50-239-1664 Supplier Waters Corp Supplier No. 186008075
This clean-up tube is designed for use in the first step of QuEChERS sample preparation. It is a d-SPE tube containing C18, specifically formulated for the analysis of acidic pesticides. QuEChERS methods are widely used for multi-pesticide residue analyses in food samples.
  • Used for extract from step 1 in QuEChERS sample preparation.
  • d-SPE tube contains C18 for acidic pesticide analysis.
  • Utilized for multi-pesticide residue analyses in food.
  • Contains 150 mg MgSO4 and 50 mg C18.
  • Tube size of 2 mL.
  • Suitable for AOAC methods.
  • Supports QuEChERS and Dispersive SPE techniques.