Learn More
Waters Corp Waters Corporation Centrifuge Tube, 50 mL, Empty, 50 pk
Catalog No. 50466503 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This empty 50 mL centrifuge tube is designed for QuEChERS (Quick, Easy, Cheap, Effective, Rugged, and Safe) extraction, a sample preparation technique for multi-residue analysis in various food and agricultural products.
  • Application: Food
  • Product type: Sample extraction
  • Packaging: Tube
  • Technique: QuEChERS, dispersive SPE
  • Volume: 50 mL
  • Quantity: 50 tubes per package
Catalog No. 50-466-503 Supplier Waters Corp Supplier No. 186006814
This empty 50 mL centrifuge tube is designed for QuEChERS (Quick, Easy, Cheap, Effective, Rugged, and Safe) extraction, a sample preparation technique for multi-residue analysis in various food and agricultural products.
  • Application: Food
  • Product type: Sample extraction
  • Packaging: Tube
  • Technique: QuEChERS, dispersive SPE
  • Volume: 50 mL
  • Quantity: 50 tubes per package