Learn More
Waters Corp QuEChERS dSPE Tube, 15 mL, MgSO4, PSA, C18, Al-N
Catalog No. 502392933 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This 15 mL tube cleans up QuEChERS samples. It contains PSA, C18, and Alumina neutral sorbents, suitable for mycotoxin and multi-mycotoxin residue analyses in grains. The kit simplifies LC analysis by streamlining extraction and cleaning, removing matrix interferences in food, environmental, pharmaceutical, and forensic applications.
  • Includes 50 x 15 mL tubes with 750 mg magnesium sulfate, 250 mg PSA, 150 mg C18, and 150 mg aluminum nitrate.
  • Uses a simple, one-step QuEChERS extraction method.
  • Magnesium sulfate precipitates proteins; PSA removes polar compounds; C18 retains hydrophobic compounds.
  • Aluminum nitrate adjusts pH.
  • Achieves high recovery of target analytes and efficient removal of matrix interferences for reliable LC analysis.
  • Compatible with diverse matrices and many analytes.
  • Reusable tubes offer cost-effective, eco-friendly sample preparation.
Catalog No. 50-239-2933 Supplier Waters Corp Supplier No. 186008080
This 15 mL tube cleans up QuEChERS samples. It contains PSA, C18, and Alumina neutral sorbents, suitable for mycotoxin and multi-mycotoxin residue analyses in grains. The kit simplifies LC analysis by streamlining extraction and cleaning, removing matrix interferences in food, environmental, pharmaceutical, and forensic applications.
  • Includes 50 x 15 mL tubes with 750 mg magnesium sulfate, 250 mg PSA, 150 mg C18, and 150 mg aluminum nitrate.
  • Uses a simple, one-step QuEChERS extraction method.
  • Magnesium sulfate precipitates proteins; PSA removes polar compounds; C18 retains hydrophobic compounds.
  • Aluminum nitrate adjusts pH.
  • Achieves high recovery of target analytes and efficient removal of matrix interferences for reliable LC analysis.
  • Compatible with diverse matrices and many analytes.
  • Reusable tubes offer cost-effective, eco-friendly sample preparation.