Learn More
Waters Corp Waters QuEChERS Tube, 900 mg MgSO4, 300 mg PSA, 300 mg C18, 15 mL
Catalog No. 502038325 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This clean-up tube is designed for the extract from step 1 in QuEChERS sample preparation. It contains PSA and C18 sorbents for effective cleanup of fatty produce and cereals. QuEChERS methods are widely used for multi-pesticide residue analyses in food.
  • Used for extract cleanup in step 1 of QuEChERS sample preparation.
  • Contains PSA and C18 sorbents for cleanup of fatty produce and cereals.
  • Suitable for multi-pesticide residue analyses in food using QuEChERS methods.
  • Supplied as a 50 pack of tubes with d-SPE components.
Catalog No. 50-203-8325 Supplier Waters Corp Supplier No. 186008078
This clean-up tube is designed for the extract from step 1 in QuEChERS sample preparation. It contains PSA and C18 sorbents for effective cleanup of fatty produce and cereals. QuEChERS methods are widely used for multi-pesticide residue analyses in food.
  • Used for extract cleanup in step 1 of QuEChERS sample preparation.
  • Contains PSA and C18 sorbents for cleanup of fatty produce and cereals.
  • Suitable for multi-pesticide residue analyses in food using QuEChERS methods.
  • Supplied as a 50 pack of tubes with d-SPE components.