Learn More
Waters Corp QuEChERS Sample Cleanup Tube, d-SPE, 15 mL
Catalog No. 502392929 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This clean-up tube is utilized for the extract from step 1 in QuEChERS sample preparation. It is a dispersive solid-phase extraction (d-SPE) tube that contains PSA, C18, and GCB for the effective clean up of fatty and pigmented samples. QuEChERS methods are employed for multi-pesticide residue analyses in food.
  • Contains 900 mg MgSO4, 450 mg PSA, 300 mg C18, and 50 mg GCB.
  • Designed for cleaning fatty and pigmented samples.
  • Suitable for multi-pesticide residue analyses in food.
  • Comes in a 15 mL tube format.
  • Provided in a convenient 50-pack.
Catalog No. 50-239-2929 Supplier Waters Corp Supplier No. 186008079
This clean-up tube is utilized for the extract from step 1 in QuEChERS sample preparation. It is a dispersive solid-phase extraction (d-SPE) tube that contains PSA, C18, and GCB for the effective clean up of fatty and pigmented samples. QuEChERS methods are employed for multi-pesticide residue analyses in food.
  • Contains 900 mg MgSO4, 450 mg PSA, 300 mg C18, and 50 mg GCB.
  • Designed for cleaning fatty and pigmented samples.
  • Suitable for multi-pesticide residue analyses in food.
  • Comes in a 15 mL tube format.
  • Provided in a convenient 50-pack.