Learn More
Perkin Elmer US LLC QuEChERS AOAC 2007.01 Method Extraction Kit - 50 mL, 50 ea.
Catalog No. 502105228
Encompass_Preferred
QuEChERS American AOAC Standard (Official Method 2007.01) Extraction kit contains 50 of each of the following, packed in ready-to-use 50 mL tubes: 6 g MgSO4 1.5 g Na Acetate
Catalog No. 50-210-5228 Supplier Perkin Elmer US LLC Supplier No. N9306900
QuEChERS American AOAC Standard (Official Method 2007.01) Extraction kit contains 50 of each of the following, packed in ready-to-use 50 mL tubes: 6 g MgSO4 1.5 g Na Acetate