Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ QuEChERS AOAC 2007.01 Method Extraction Kits

Catalog No. 03254137 Shop All Thermo Scientific Products
Change view
Click to view available options
Includes:
Salts in Pouches
Salts in Pouches, Tubes
Salts in Pouches, Tubes, Ceramic Homogenizers
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
03-254-137 Salts in Pouches, Tubes
03-254-136 Salts in Pouches, Tubes, Ceramic Homogenizers
03-254-180 Salts in Pouches
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 03-254-137 Supplier Thermo Scientific™ Supplier No. S115AOACKIT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Use Thermo Scientific™ QuEChERS AOAC 2007.01 Method Extraction Kits to prepare complex samples for LC-MS or GC-MS analysis of residues, toxins, and other contaminants.

Use Thermo Scientific™ QuEChERS Extraction Kits to prepare complex sample matrices such as fruits and vegetables for LC-MS or GC-MS analysis of pesticide residues, veterinary drug residues, and toxins using the AOAC 2007.01 Method. Our QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) kits contain pre-packaged, ready-weighed salts, sorbents, and buffers designed to streamline your workflow, provide excellent recoveries for a wide variety of analytes, minimize the potential for error, save time and money, and deliver high-quality, reproducible results.

QuEChERS AOAC 2007.01 Method Extraction Kits

Extract analytes of interest from a homogenized sample using extraction salts. Use these kits with 10 g or 15 g samples. Choose from:

  • Extraction Salts in Pouches, No Tubes
  • Extraction Salts in Pouches with 50 mL Tubes
  • Extraction Salts in Pouches with 50 mL Tubes and Ceramic Homogenizers

Chemicals used in the AOAC 2007.01 Method

  • Acetonitrile: used to perform single-phase extraction on a sample
  • Anhydrous MgSO4: induces phase separation between water content in sample and acetonitrile layer during liquid-liquid partitioning; removes remaining water from sample during dSPE
  • NaOAc: buffers the sample to stabilize pH
  • PSA: primary secondary amine, used to remove polar matrix components, citric co-extractives, and free fatty acids from sample during dispersive solid phase extraction
  • C18: removes fats, sterols, and other non-polar interferences from sample
  • GCB: graphitized carbon black, removes pigment (not recommended for use with planar pesticides)
TRUSTED_SUSTAINABILITY

Specifications

Description QuEChERS AOAC Extraction Kit for 15 g sample; 6 g Anhydrous MgSO4, 1.5 g NaOAc in Pouches; Empty 50 mL Tubes; 50/Pk.
Type QuEChERS Extraction Kit
Includes Salts in Pouches, Tubes
Method QuEChERS sample preparation for AOAC 2007.01 Method
Salt Composition 6 g Anhydrous MgSO4, 1.5 g NaOAc
Sample Size 15 g sample
Tube Size 50 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.