Learn More
Perkin Elmer US LLC QuEChERS Fruit and Vegetables EN 15662 Clean-Up Kit - 15 mL, 50 ea.
Catalog No. 502105241
Encompass_Preferred
QuEChERS EN15662 Method Clean-up Kit for Fruits and Vegetables contains 50 of each of the following, packed in ready-to-use 15 mL tubes: 900 mg MgSO4 - to remove excess water. 150 mg PSA - to remove sugars, fatty acids, organic acids, and anthocyanine pigments.
Catalog No. 50-210-5241 Supplier Perkin Elmer US LLC Supplier No. N9306921
QuEChERS EN15662 Method Clean-up Kit for Fruits and Vegetables contains 50 of each of the following, packed in ready-to-use 15 mL tubes: 900 mg MgSO4 - to remove excess water. 150 mg PSA - to remove sugars, fatty acids, organic acids, and anthocyanine pigments.