Learn More
Perkin Elmer US LLC QuEChERS Method EN 15662 SPE Powder Pack, 500 ea.
Catalog No. 502113659
Encompass_Preferred
QuEChERS European Standard (EN 15662) Method Powder Pack contains 500 of each of the following: 4 g MgSO4 1 g Na Citrate 0.5 g Na Citrate Sesquihydrate 1 g NaCl This Powder Pack can also be found as a part of the QuEChERS EN 15662 Method Extraction Kit (N9306901).
Catalog No. 50-211-3659 Supplier Perkin Elmer US LLC Supplier No. N9306940
QuEChERS European Standard (EN 15662) Method Powder Pack contains 500 of each of the following: 4 g MgSO4 1 g Na Citrate 0.5 g Na Citrate Sesquihydrate 1 g NaCl This Powder Pack can also be found as a part of the QuEChERS EN 15662 Method Extraction Kit (N9306901).