Learn More
MilliporeSigma™ Supelco™ QuEChERS Shaker and Rack Starter Kit
Catalog No. 111005859 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Description:
AC/DC input, Schuko plug
AC/DC input, USA compatible plug
Voltage:
115 V
230 V
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Voltage
11-100-5859 AC/DC input, Schuko plug 230 V
11-100-5858 AC/DC input, USA compatible plug 115 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-5859 Supplier MilliporeSigma™ Supelco™ Supplier No. 55438U
Add to Cart
Add to Cart
The QuEChERS Shaker and Rack Starter Kit includes one each of the following:. Benchmixer XL multi-tube vortexer (Z765503 or Z765511). 50 mL QuEChERS Extraction Tube Shaker Rack (55279-U). 15 mL QuEChERS Cleanup Tube Shaker Rack (Z765589). 2 mL QuEChERS Cleanup Tube Shaker Rack (Z765554).
The Benchmixer XL is a heavy duty vortex mixer designed to facilitate hands-free mixing in tubes, flasks, vials or cylinders. It is ideal for the QuEChERS pesticide analysis method and is instantly adaptable to an exceptionally broad range of vessels, racks and applications (horizontal or vertical).The precisely balanced drive system includes an oversized motor that delivers accurate, reproducible speeds up to 2500 rpm with rattle-free operation and minimal vibration. The programmable pulsing function allows for time specific interval mixing, increasing agitation.The 7 x 12 inch platform accommodates a wide variety of standard, pre-drilled tube racks (a complete listing can be found in the ordering information section). Additionally, BenchMixer XL can easily be adapted to almost any type of vessel commonly found in the laboratory (please feel free to contact us for details).Using BenchMixer XL is easy; after loading the sample rack and tightening the locking knobs, select the desired speed and run time on the large digital control panel. Then, simply press 'Start' and the BenchMixer XL will perform unattended mixing for up to 100 hours.BenchMixer XL's large capacity and adaptability make it an ideal choice for the medium to high throughput laboratory. Hands-free, high capacity vortexing. Up to 2500 rpm, a true vortex in every tube. Assortment of accessories for a variety of tubes, flasks & plates. Pulsing program for interval mixing.
TRUSTED_SUSTAINABILITY
Specifications
Description AC/DC input, Schuko plug
For Use With (Application) Food and Beverages
Type SPE Accessory
Quantity 1/Pk.
Voltage 230 V