Learn More
TA Instruments Quench Cooling Accessory (QCA), Temperature Range-180°C to 550°C, -130°C for DSC 25P, Requires Additional Manual Lids and Covers, Compatible with Discovery DSCs
Catalog No. 5000580690 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Quench Cooling Accessory (QCA) enables cooling from -180°C to 550°C for Discovery DSC series. DSC 25P supports down to -130°C. For quench cooling of autosampler-enabled Discovery DSCs, the following parts are also required: P/N: 973061.903 Manual Insulated Cover and 973252.001 Manual Silver Lid.
Catalog No. 50-005-80690 Supplier TA Instruments Supplier No. 970347.901
Waters TA Instruments Quench Cooling Accessory (QCA) enables cooling from -180°C to 550°C for Discovery DSC series. DSC 25P supports down to -130°C. For quench cooling of autosampler-enabled Discovery DSCs, the following parts are also required: P/N: 973061.903 Manual Insulated Cover and 973252.001 Manual Silver Lid.