Learn More
TA Instruments Quench Cooling Can, Designed for Finned Air Cooling System (FACS), Rapid Cooling, DSC Cell from 725°C to Ambient, Customer Supplied Cooling Medium, Suitable for Tzero Pressure DSC Cell
Catalog No. 5000580711 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Quench Cooling Can is designed to supplement the cooling performance of the Finned Air Cooling System (FACS). It provides rapid cooling of the DSC cell from 725°C to ambient temperature using a customer supplied cooling medium such as ice, solid CO2, or liquid nitrogen. This accessory does not extend the temperature range of the FACS to sub-ambient temperatures. It is also suitable for use with the Tzero Pressure DSC Cell (P/N: 970900.901).
Catalog No. 50-005-80711 Supplier TA Instruments Supplier No. 970036.901
Waters TA Instruments Quench Cooling Can is designed to supplement the cooling performance of the Finned Air Cooling System (FACS). It provides rapid cooling of the DSC cell from 725°C to ambient temperature using a customer supplied cooling medium such as ice, solid CO2, or liquid nitrogen. This accessory does not extend the temperature range of the FACS to sub-ambient temperatures. It is also suitable for use with the Tzero Pressure DSC Cell (P/N: 970900.901).