Learn More
Selleck Chemical LLC Quercetin 100mg 117-39-5 Sophoretin
SDP
Supplier:  Selleck Chemical LLC S2391100MG
Encompass_Preferred
Quercetin (Sophoretin), a natural flavonoid present in vegetables, fruit and wine, is a stimulator of recombinant SIRT1 and also a PI3K inhibitor with IC50 of 2.4-5.4 ?M. Quercetin induces mitophagy, apoptosis and protective autophagy. Phase 4. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2357