Learn More
Medchemexpress LLC Quercetin 3,3'-dimethyl ether | 4382-17-6 | 99.0% | 330.29 | 1 MG
SDP
Catalog No. 5000425857
Encompass_Preferred
Quercetin 3,3'-dimethyl ether is a compound with demonstrated antioxidant activity, intended for research use only. It has been observed to inhibit procoagulant activity in human monocytes and exhibits cytotoxicity against mouse P388 cells.
  • Possesses antioxidant activity
  • Inhibits procoagulant activity in monocytes (IC50 of 3.9 x 10-6 M)
  • Counteracts IL1-induced tissue factor expression
  • Exhibits cytotoxicity against mouse P388 cells (ED50 of 1.7 μg/mL)
  • Suitable for research applications
Catalog No. 50-004-25857 Supplier Medchemexpress LLC Supplier No. HYN91351MG
Quercetin 3,3'-dimethyl ether is a compound with demonstrated antioxidant activity, intended for research use only. It has been observed to inhibit procoagulant activity in human monocytes and exhibits cytotoxicity against mouse P388 cells.
  • Possesses antioxidant activity
  • Inhibits procoagulant activity in monocytes (IC50 of 3.9 x 10-6 M)
  • Counteracts IL1-induced tissue factor expression
  • Exhibits cytotoxicity against mouse P388 cells (ED50 of 1.7 μg/mL)
  • Suitable for research applications