Learn More
Apexbio Technology LLC QUERCETIN 3-4-7 TRIME-5mg
SDP
Catalog No. 5000897652
Encompass_Preferred
Quercetin 3 4 7-trimethyl ether (CAS No 6068-80-0) is a flavonoid compound investigated in the biomedical research category for its potential therapeutic properties Originating from various plant sources this compound is notable for its antioxidant and anti-inflammatory activities Its mechanism involves modulating cellular pathways like oxidative stress markers and inflammatory mediators impacting enzymes and receptors involved in these processes In vitro activity typically falls within the nanomolar to low micromolar range demonstrating significant efficacy at these concentrations Common applications include studies with cell models of cancer and inflammation as well as animal studies to explore its pharmacological effects It is also used in drug screening to evaluate its potential as a therapeutic agent Experimental concentrations typically depend on the specific research design ensuring tailored approaches for different study needs
Catalog No. 50-008-97652 Supplier Apexbio Technology LLC Supplier No. N28655
Quercetin 3 4 7-trimethyl ether (CAS No 6068-80-0) is a flavonoid compound investigated in the biomedical research category for its potential therapeutic properties Originating from various plant sources this compound is notable for its antioxidant and anti-inflammatory activities Its mechanism involves modulating cellular pathways like oxidative stress markers and inflammatory mediators impacting enzymes and receptors involved in these processes In vitro activity typically falls within the nanomolar to low micromolar range demonstrating significant efficacy at these concentrations Common applications include studies with cell models of cancer and inflammation as well as animal studies to explore its pharmacological effects It is also used in drug screening to evaluate its potential as a therapeutic agent Experimental concentrations typically depend on the specific research design ensuring tailored approaches for different study needs