Learn More
Cayman Chemical Quercetin 3-D-galactoside, Purity: ≥98%, CAS: 482-36-0, 5 mg
SDP
Catalog No. 502966282
Encompass_Preferred
A natural 3-O-galactoside of quercetin that has powerful antioxidant action through its ability to scavenge free radicals (IC50s = 3.54 and 5.44 µg/ml in ABTS and DPPH assays, respectively)
Catalog No. 50-296-6282 Supplier Cayman Chemical Supplier No. 186485 MG
A natural 3-O-galactoside of quercetin that has powerful antioxidant action through its ability to scavenge free radicals (IC50s = 3.54 and 5.44 µg/ml in ABTS and DPPH assays, respectively)