Learn More
Sigma Aldrich Fine Chemicals Biosciences Quercetin 3 glucoside phyp20MG
Catalog No. 502902629
Encompass
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity water residual solvents inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH Co. KG
Catalog No. 50-290-2629 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. PHL8923020MG
Add to Cart
Add to Cart
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity water residual solvents inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH Co. KG