Learn More
Medchemexpress LLC Quercetin 3-O-(6''-O-malonyl)-β-D-glucoside | 96862-01-0 | 99.65% | 550.42 | 5 MG
SDP
Catalog No. 5000425387
Encompass_Preferred
Quercetin 3-O-(6''-O-malonyl)-β-D-glucoside is a flavonol glycoside that possesses antioxidant activity. In a cell-free environment, it exhibits excellent antioxidant ability and protects pBR322 DNA from oxidative damage.
  • Exhibits excellent antioxidant ability in a cell-free environment.
  • Protects pBR322 DNA from oxidative damage.
Catalog No. 50-004-25387 Supplier Medchemexpress LLC Supplier No. HYN93975MG
Quercetin 3-O-(6''-O-malonyl)-β-D-glucoside is a flavonol glycoside that possesses antioxidant activity. In a cell-free environment, it exhibits excellent antioxidant ability and protects pBR322 DNA from oxidative damage.
  • Exhibits excellent antioxidant ability in a cell-free environment.
  • Protects pBR322 DNA from oxidative damage.