Learn More
Medchemexpress LLC Quercetin-3-O-D-glucosyl]-(1-2)-L-rhamnoside | 143016-74-4 | 10 MG
SDP
Catalog No. 5000418791
Encompass_Preferred
Quercetin-3-O-D-glucosyl]-(1-2)-L-rhamnoside is a flavonoid glycoside with antioxidant activity and free radical scavenging activity. It effectively scavenges free radicals, including ABTS+, making it valuable for research into antioxidant-related diseases such as cardiovascular and neurodegenerative conditions.
  • A flavonoid glycoside with antioxidant properties
  • Scavenges free radicals like ABTS+
  • Useful in the study of cardiovascular and neurodegenerative diseases
  • Classified as a flavonoid, flavonol, phenol, and polyphenol
  • Naturally sourced from plants such as Ginkgo biloba
Catalog No. 50-004-18791 Supplier Medchemexpress LLC Supplier No. HYN760710MG
Quercetin-3-O-D-glucosyl]-(1-2)-L-rhamnoside is a flavonoid glycoside with antioxidant activity and free radical scavenging activity. It effectively scavenges free radicals, including ABTS+, making it valuable for research into antioxidant-related diseases such as cardiovascular and neurodegenerative conditions.
  • A flavonoid glycoside with antioxidant properties
  • Scavenges free radicals like ABTS+
  • Useful in the study of cardiovascular and neurodegenerative diseases
  • Classified as a flavonoid, flavonol, phenol, and polyphenol
  • Naturally sourced from plants such as Ginkgo biloba