Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Quercetin-3-O-D-glucosyl]-(1-2)-L-rhamnoside | 143016-74-4 | 5 MG
SDP

Catalog No. 5000419956
Encompass_Preferred

Quercetin-3-O-D-glucosyl]-(1-2)-L-rhamnoside is a flavonoid glycoside with strong antioxidant activity and free radical scavenging capabilities. This compound effectively scavenges free radicals, such as ABTS+, making it valuable for research into antioxidant-related conditions, including cardiovascular and neurodegenerative diseases.

  • High purity of 99.87%.
  • Exhibits concentration-dependent targeted ABTS+ radical scavenging activity.
  • Derived from plants, including Ginkgo biloba.
  • Molecular weight of 610.52.
  • Chemical formula C27H30O16.
  • Appears as a white to yellow solid.
  • Soluble in DMSO (100 mg/mL) and H2O (14.29 mg/mL).
  • Store at 4°C, protected from light.
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light.

Catalog No. 50-004-19956 Supplier Medchemexpress LLC Supplier No. HYN76075MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Quercetin-3-O-D-glucosyl]-(1-2)-L-rhamnoside is a flavonoid glycoside with strong antioxidant activity and free radical scavenging capabilities. This compound effectively scavenges free radicals, such as ABTS+, making it valuable for research into antioxidant-related conditions, including cardiovascular and neurodegenerative diseases.

  • High purity of 99.87%.
  • Exhibits concentration-dependent targeted ABTS+ radical scavenging activity.
  • Derived from plants, including Ginkgo biloba.
  • Molecular weight of 610.52.
  • Chemical formula C27H30O16.
  • Appears as a white to yellow solid.
  • Soluble in DMSO (100 mg/mL) and H2O (14.29 mg/mL).
  • Store at 4°C, protected from light.
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.